siRNA Details
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253783.1 | NP_001240712.1 | 12 | 1251 | ||
NM_001253784.1 | NP_001240713.1 | 11 | 1229 | |||
NM_001281803.1 | NP_001268732.1 | 10 | 1233 | |||
NM_054046.5 | NP_473387.1 | 10 | 1142 | |||
GenBank mRNA | AK028726.1 | - | 11 | 1201 | ||
AK034028.1 | - | 1108 | ||||
AK046271.1 | - | 10 | 1102 | |||
AK050405.1 | - | 11 | 1200 | |||
AK081295.1 | - | 10 | 1132 | |||
AK086174.1 | - | 10 | 1105 | |||
AK087778.1 | - | 10 | 1113 | |||
AK129019.1 | - | 10 | 1184 | |||
AK133979.1 | - | 10 | 1116 | |||
AK150334.1 | - | 10 | 1078 | |||
AK150994.1 | - | 10 | 1078 | |||
AK164147.1 | - | 10 | 1114 | |||
BC003306.1 | - | 12 | 1202 | 3154712 | ||
CN705834.1 | - | 3 | 183 | |||
X96708.1 | - | 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
