siRNA Details
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110327.1 | NP_001103797.1 | 10 | 1027 | ||
NM_011806.3 | NP_035936.3 | 10 | 1027 | |||
GenBank mRNA | AK029147.1 | - | 10 | 969 | ||
AK031887.1 | - | 9 | 845 | |||
AK039563.1 | - | 10 | 995 | |||
AK046201.1 | - | 10 | 1098 | |||
AK078015.1 | - | 10 | 930 | |||
AK145803.1 | - | 8 | 2936 | |||
AK160595.1 | - | 10 | 1000 | |||
BC045141.1 | - | 9 | 847 | 6307203 | ||
BC060109.1 | - | 10 | 949 | |||
U70017.1 | - | 10 | 982 |
