siRNA Details
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110327.1 | NP_001103797.1 | 6 | 636 | ||
NM_011806.3 | NP_035936.3 | 6 | 636 | |||
GenBank mRNA | AK029147.1 | - | 6 | 578 | ||
AK031887.1 | - | 5 | 454 | |||
AK039563.1 | - | 6 | 604 | |||
AK046201.1 | - | 6 | 532 | |||
AK078015.1 | - | 6 | 539 | |||
AK145803.1 | - | 6 | 598 | |||
AK160595.1 | - | 6 | 609 | |||
BC045141.1 | - | 5 | 456 | 6307203 | ||
BC060109.1 | - | 6 | 559 | |||
U70017.1 | - | 6 | 591 |
