siRNA Details
Gene Symbol: Dmtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110327.1 | NP_001103797.1 | 10 | 1016 | ||
NM_011806.3 | NP_035936.3 | 10 | 1016 | |||
GenBank mRNA | AK029147.1 | - | 10 | 958 | ||
AK031887.1 | - | 9 | 834 | |||
AK039563.1 | - | 10 | 984 | |||
AK046201.1 | - | 10 | 1087 | |||
AK078015.1 | - | 10 | 919 | |||
AK145803.1 | - | 8 | 2925 | |||
AK160595.1 | - | 10 | 989 | |||
BC045141.1 | - | 9 | 836 | 6307203 | ||
BC060109.1 | - | 10 | 938 | |||
U70017.1 | - | 10 | 971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
