siRNA Details
Gene Symbol: Fiz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110328.1 | NP_001103798.1 | 2 | 310 | ||
NM_001110329.1 | NP_001103799.1 | 2 | 253 | |||
NM_001110330.1 | NP_001103800.1 | 2 | 183 | |||
NM_011813.3 | NP_035943.3 | 2 | 294 | |||
GenBank mRNA | AF126746.1 | - | 2 | 244 | ||
AF126747.1 | - | 2 | 252 | |||
AK135672.1 | - | 2 | 183 | |||
AK139120.1 | - | 2 | 267 | |||
AK142205.1 | - | 2 | 203 | |||
AK162454.1 | - | 2 | 175 | |||
AK170633.1 | - | 2 | 179 | |||
AK196020.1 | - | 1 | 129 | |||
AK203569.1 | - | 1 | 130 | |||
AK220212.1 | - | 2 | 183 | |||
BC006633.1 | - | 2 | 294 | 3482191 | ||
BC106113.1 | - | 2 | 169 | 6492060 | ||
BC138231.1 | - | 2 | 244 | 8861250 | ||
BC138232.1 | - | 2 | 269 | 8861251 | ||
BC145207.1 | - | 2 | 269 | 9053758 | ||
BU053482.1 | - | 2 | 585 | |||
BY344494.1 | - | 2 | 287 | |||
CJ131593.1 | - | 2 | 252 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
