siRNA Details
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011814.2 | NP_035944.2 | 17 | 2924 | ||
GenBank mRNA | AK154243.1 | - | 17 | 2923 | ||
AK192714.1 | - | 1 | 264 | |||
AK195317.1 | - | 1 | 264 | |||
BC057007.1 | - | 17 | 2819 | 6816943 | ||
BC062971.1 | - | 17 | 2924 | 6415619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
