siRNA Details
Gene Symbol: Fxr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011814.2 | NP_035944.2 | 16 | 2255 | ||
GenBank mRNA | AB025269.1 | - | 15 | 1819 | ||
AK154243.1 | - | 16 | 2254 | |||
AK212955.1 | - | 1 | 30 | |||
BC057007.1 | - | 16 | 2150 | 6816943 | ||
BC062971.1 | - | 16 | 2255 | 6415619 |

More Information
Other Species Targeted

Gene Ontology Categories:
