siRNA Details
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278269.1 | NP_001265198.1 | 14 | 2220 | ||
NM_011815.5 | NP_035945.1 | 15 | 2358 | |||
GenBank mRNA | AF001863.1 | - | 14 | 2196 | ||
AF061744.1 | - | 15 | 2334 | |||
AK038225.1 | - | 14 | 2248 | |||
AK137907.1 | - | 14 | 2176 | |||
AK172164.1 | - | 8 | 590 | |||
BC141352.1 | - | 15 | 2299 | 9055964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
