siRNA Details
Gene Symbol: Fyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278269.1 | NP_001265198.1 | 10 | 1945 | ||
NM_011815.5 | NP_035945.1 | 10 | 1945 | |||
GenBank mRNA | AF001863.1 | - | 10 | 1921 | ||
AF061744.1 | - | 10 | 1921 | |||
AK038161.1 | - | 1 | 1403 | |||
AK038225.1 | - | 10 | 1973 | |||
AK046759.1 | - | 2 | 91 | |||
AK137907.1 | - | 10 | 1901 | |||
AK172164.1 | - | 3 | 177 | |||
BC141352.1 | - | 10 | 1886 | 9055964 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
