siRNA Details
Gene Symbol: Grem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011824.4 | NP_035954.1 | 2 | 424 | ||
GenBank mRNA | AF045801.1 | - | 1 | 265 | ||
AF108189.1 | - | 1 | 334 | |||
AK135022.1 | - | 2 | 459 | |||
AK157793.1 | - | 2 | 425 | |||
AK164871.1 | - | 2 | 425 | |||
BC015293.1 | - | 2 | 394 | 4216669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
