siRNA Details
Gene Symbol: Grem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011825.1 | NP_035955.1 | 2 | 715 | ||
GenBank mRNA | AB011030.1 | - | 2 | 715 | ||
AK142008.1 | - | 2 | 717 | |||
AK161827.1 | - | 2 | 695 | |||
BC079905.1 | - | 2 | 696 | 30645267 |
