siRNA Details
Gene Symbol: Hax1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011826.4 | NP_035956.1 | 3 | 540 | ||
GenBank mRNA | AF023482.1 | - | 3 | 512 | ||
AF023483.1 | - | 2 | 675 | |||
AK002256.1 | - | 3 | 491 | |||
AK010633.1 | - | 3 | 524 | |||
AK088746.1 | - | 3 | 522 | |||
BC006688.1 | - | 3 | 469 | 3582652 | ||
BC098225.1 | - | 3 | 494 | 1494110 | ||
BC147481.1 | - | 3 | 471 | 9056226 |

More Information
Other Species Targeted

Gene Ontology Categories:
