siRNA Details
Gene Symbol: Hcst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011827.3 | NP_035957.2 | 4 | 426 | ||
GenBank mRNA | AF172930.1 | - | 4 | 426 | ||
AK008005.1 | - | 4 | 425 | |||
BC069220.1 | - | 350 | 640698 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
