siRNA Details
Gene Symbol: Hs2st1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011828.3 | NP_035958.3 | 6 | 1084 | ||
GenBank mRNA | AF060178.1 | - | 6 | 762 | ||
AF169243.1 | - | 6 | 751 | |||
AK088698.1 | - | 6 | 1060 | |||
AK165112.1 | - | 6 | 1050 | |||
BC025443.1 | - | 6 | 1012 | 5068428 | ||
BC059008.1 | - | 6 | 841 | 6849136 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
