siRNA Details
Gene Symbol: Mapk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291033.1 | NP_001277962.1 | 4 | 861 | ||
NM_001291034.1 | NP_001277963.1 | 3 | 936 | |||
NM_001291035.1 | NP_001277964.1 | 3 | 1077 | |||
NM_001291036.1 | NP_001277965.1 | 4 | 926 | |||
NM_001291037.1 | NP_001277966.1 | 3 | 1175 | |||
NM_011841.2 | NP_035971.1 | 4 | 811 | |||
GenBank mRNA | AB019373.1 | - | 4 | 761 | ||
AF126159.1 | - | 3 | 574 | |||
AF126160.1 | - | 3 | 624 | |||
AF126161.1 | - | 3 | 994 | |||
AK148119.1 | - | 4 | 793 | |||
AK155187.1 | - | 4 | 729 | |||
AY534740.1 | - | 4 | 745 | |||
BC013745.1 | - | 3 | 1094 | |||
BC033598.1 | - | 4 | 901 | |||
BC051020.1 | - | 2 | 1455 | |||
BC058096.1 | - | 2 | 1455 | |||
BC082315.1 | - | 3 | 998 | |||
BC100398.1 | - | 3 | 878 | 30470655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
