siRNA Details
Gene Symbol: Mta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011842.3 | NP_035972.3 | 2, 3 | 527 | ||
GenBank mRNA | AF159259.1 | - | 2, 3 | 402 | ||
AK077548.1 | - | 2, 3 | 509 | |||
AK088158.1 | - | 2, 3 | 440 | |||
AK147099.1 | - | 2, 3 | 506 | |||
AK149834.1 | - | 2, 3 | 573 | |||
AK157717.1 | - | 2, 3 | 431 | |||
AK159979.1 | - | 2, 3 | 503 | |||
AK219501.1 | - | 1, 2 | 60 | |||
BC079847.1 | - | 2, 3 | 527 | 6419411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
