siRNA Details
Gene Symbol: Mmp17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011846.4 | NP_035976.3 | 8, 9 | 1306 | ||
GenBank mRNA | AB021224.2 | - | 8, 9 | 1312 | ||
AJ010731.1 | - | 8, 9 | 1310 | |||
AK079221.1 | - | 1078 | ||||
AK143521.1 | - | 8, 9 | 1306 | |||
BC051917.1 | - | 8, 9 | 1306 | 5716458 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
