siRNA Details
Gene Symbol: Nek4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF099067.1 | - | 13 | 2205 | ||
AJ223071.1 | - | 13 | 2295 | |||
AK078809.1 | - | 12 | 2172 | |||
AK132803.1 | - | 12 | 2089 | |||
AK147381.1 | - | 10 | 5992 | |||
AK149147.1 | - | 4 | 373 | |||
AK149584.1 | - | 12 | 2089 | |||
BC057939.1 | - | 13 | 2207 | 5325558 | ||
Y09234.1 | - | 12 | 2151 |
