siRNA Details
Gene Symbol: Oas1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF480417.1 | - | 3 | 533 | ||
BC018470.1 | - | 517 | 3493001 | |||
BC043339.1 | - | 3 | 548 | 5004267 | ||
X58077.1 | - | 460 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
