siRNA Details
Gene Symbol: Tenm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290702.1 | NP_001277631.1 | 9 | 1803 | ||
NM_011856.4 | NP_035986.3 | 10 | 1926 | |||
GenBank mRNA | AB025411.1 | - | 10 | 1963 | ||
AJ245710.1 | - | 2 | 138 | |||
AK031198.1 | - | 9 | 1627 | |||
BC141361.1 | - | 9 | 1804 | 9055973 |
