siRNA Details
Gene Symbol: Pacsin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286743.1 | NP_001273672.1 | 6 | 1065 | ||
NM_001286744.1 | NP_001273673.1 | 6 | 971 | |||
NM_011861.3 | NP_035991.1 | 6 | 1030 | |||
NM_178365.4 | NP_848142.1 | 6 | 1048 | |||
GenBank mRNA | AK043967.1 | - | 6 | 1034 | ||
AK077958.1 | - | 1 | 150 | |||
AK158686.1 | - | 6 | 1068 | |||
BC014698.1 | - | 6 | 919 | 4527708 | ||
X85124.1 | - | 6 | 939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
