siRNA Details
Gene Symbol: Papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289477.1 | NP_001276406.1 | 9, 10 | 1248 | ||
NM_001289478.1 | NP_001276407.1 | 9, 10 | 1435 | |||
NM_001289479.1 | NP_001276408.1 | 10, 11 | 1546 | |||
NM_011863.2 | NP_035993.1 | 8, 9 | 1258 | |||
GenBank mRNA | AK146507.1 | - | 8, 9 | 1131 | ||
AK153296.1 | - | 8, 9 | 1128 | |||
AK160875.1 | - | 10, 11 | 1450 | |||
AK168982.1 | - | 8, 9 | 1131 | |||
AK169117.1 | - | 9, 10 | 1310 | |||
AK169124.1 | - | 8, 9 | 1125 | |||
AK169204.1 | - | 9, 10 | 1310 | |||
AK211888.1 | - | 1, 2 | 52 | |||
BC066055.1 | - | 9, 10 | 1314 | 6833756 | ||
U34883.1 | - | 8, 9 | 1135 |
