siRNA Details
Gene Symbol: Dazap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011873.2 | NP_036003.2 | 2 | 132 | ||
GenBank mRNA | AF085348.1 | - | 2 | 97 | ||
AK002595.1 | - | 2 | 169 | |||
AK050159.1 | - | 2 | 146 | |||
AK088916.1 | - | 2 | 154 | |||
AK132139.1 | - | 2 | 146 | |||
AK137246.1 | - | 2 | 133 | |||
AK145398.1 | - | 2 | 166 | |||
AK146970.1 | - | 2 | 157 | |||
AK151730.1 | - | 1 | 848 | |||
AK152511.1 | - | 1 | 848 | |||
AK169769.1 | - | 1 | 432 | |||
BC014808.1 | - | 2 | 132 | 4506124 | ||
BC029351.1 | - | 1 | 63 | |||
BC083347.1 | - | 2 | 124 | 6829350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
