siRNA Details
Gene Symbol: Psmc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011874.2 | NP_036004.2 | 11 | 1283 | ||
GenBank mRNA | AK050677.1 | - | 11 | 1261 | ||
AK077507.1 | - | 11 | 1246 | |||
AK153991.1 | - | 11 | 1266 | |||
AK160644.1 | - | 11 | 1245 | |||
AK160718.1 | - | 11 | 1237 | |||
AK167041.1 | - | 11 | 1228 | |||
AK167528.1 | - | 11 | 1238 | |||
AK169224.1 | - | 11 | 1246 | |||
AK169244.1 | - | 11 | 1745 | |||
BC012411.1 | - | 3 | 505 | |||
BC012708.1 | - | 11 | 1244 | 4211677 | ||
BC092265.1 | - | 11 | 1227 | 3671250 | ||
BC094063.1 | - | 6 | 601 | |||
L76223.1 | - | 11 | 1234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
