siRNA Details
Gene Symbol: Rngtt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF025653.1 | - | 13 | 1441 | ||
AF034568.1 | - | 13 | 1495 | |||
AK002922.1 | - | 13 | 1544 | |||
AK142016.1 | - | 13 | 1538 | |||
AK150497.1 | - | 13 | 1548 | |||
AK151463.1 | - | 13 | 1562 | |||
AK160007.1 | - | 13 | 1550 | |||
AK160706.1 | - | 13 | 1528 | |||
AK169305.1 | - | 13 | 1514 | |||
BC043657.1 | - | 13 | 1513 | 5343539 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
