siRNA Details
Gene Symbol: Sept3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011889.2 | NP_036019.2 | 5 | 563 | ||
GenBank mRNA | AF104411.1 | - | 5 | 613 | ||
AK030668.1 | - | 5 | 531 | |||
AK046722.1 | - | 5 | 656 | |||
AK141915.1 | - | 5 | 490 | |||
AK164861.1 | - | 5 | 660 | |||
AK169916.1 | - | 5 | 684 | |||
AK220520.1 | - | 3 | 362 | |||
BC055738.1 | - | 5 | 492 | 6413697 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
