siRNA Details
Gene Symbol: Sigirr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023059.3 | NP_075546.2 | 6 | 1087 | ||
GenBank mRNA | AF113795.1 | - | 5 | 604 | ||
AF239957.1 | - | 5 | 598 | |||
AK133518.1 | - | 6 | 712 | |||
AK145889.1 | - | 6 | 1090 | |||
BC010806.1 | - | 5 | 609 | 4240425 | ||
BC094069.1 | - | 6 | 693 | 4974962 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
