siRNA Details
Gene Symbol: Slc35a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011895.3 | NP_036025.2 | 7 | 1052 | ||
GenBank mRNA | AK144170.1 | - | 7 | 1045 | ||
BC012252.1 | - | 7 | 967 | 4190458 | ||
Z71268.1 | - | 7 | 873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
