siRNA Details
Gene Symbol: Sufu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025391.2 | NP_001020562.1 | 4 | 709 | ||
NM_015752.3 | NP_056567.2 | 4 | 709 | |||
GenBank mRNA | AF134893.1 | - | 4 | 649 | ||
AJ131692.1 | - | 4 | 710 | |||
AJ308625.1 | - | 4 | 388 | |||
AJ308626.1 | - | 4 | 643 | |||
AK015885.1 | - | 3 | 467 | |||
AK016976.1 | - | 4 | 688 | |||
AK047603.1 | - | 4 | 579 | |||
AK156410.1 | - | 4 | 671 | |||
BC048168.1 | - | 4 | 682 | 6821477 | ||
BC056997.1 | - | 4 | 682 | 6821477 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
