siRNA Details
Gene Symbol: Synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025292.6 | NP_079568.1 | 2 | 214 | ||
GenBank mRNA | AK008054.1 | - | 2 | 197 | ||
AK008254.1 | - | 2 | 184 | |||
AK013474.1 | - | 2 | 176 | |||
AK033514.1 | - | 2 | 215 | |||
AK163362.1 | - | 2 | 182 | |||
AY071903.1 | - | 1 | 487 | |||
AY138960.1 | - | 2 | 154 | |||
AY566156.1 | - | 2 | 187 | |||
BC027433.1 | - | 2 | 176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
