siRNA Details
Gene Symbol: Synj2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025292.6 | NP_079568.1 | 2 | 258 | ||
GenBank mRNA | AK008054.1 | - | 2 | 241 | ||
AK008254.1 | - | 2 | 228 | |||
AK013474.1 | - | 2 | 220 | |||
AK033514.1 | - | 2 | 259 | |||
AK163362.1 | - | 2 | 226 | |||
AY071903.1 | - | 1 | 531 | |||
AY138960.1 | - | 2 | 198 | |||
AY566156.1 | - | 2 | 231 | |||
BC027433.1 | - | 2 | 220 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
