siRNA Details
Gene Symbol: Taf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175770.4 | NP_786964.1 | 2 | 692 | ||
GenBank mRNA | AF144562.1 | - | 2 | 639 | ||
AK029380.1 | - | 2 | 654 | |||
AK075700.1 | - | 2 | 640 | |||
AK076362.1 | - | 2 | 655 | |||
BC029673.1 | - | 2 | 623 | 5373242 |

More Information
Other Species Targeted

Gene Ontology Categories:
