siRNA Details
Gene Symbol: Zfp68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044747.2 | NP_001038212.1 | 5 | 852 | ||
NM_001163797.1 | NP_001157269.1 | 6 | 942 | |||
NM_013844.2 | NP_038872.1 | 6 | 948 | |||
GenBank mRNA | AB024005.1 | - | 6 | 932 | ||
AK013868.1 | - | 6 | 943 | |||
AK044831.1 | - | 5 | 822 | |||
AK146092.1 | - | 6 | 909 | |||
AK153887.1 | - | 6 | 884 | |||
BC018171.1 | - | 6 | 931 | 3989444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
