siRNA Details
Gene Symbol: Ing1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB221554.1 | - | 2 | 466 | ||
AF149820.1 | - | 2 | 460 | |||
AF177755.1 | - | 2 | 448 | |||
AF177756.1 | - | 2 | 433 | |||
AF177757.1 | - | 2 | 1306 | |||
AK035490.1 | - | 2 | 463 | |||
AK076482.1 | - | 2 | 356 | |||
AK141466.1 | - | 2 | 356 | |||
AK145553.1 | - | 2 | 358 | |||
AK157424.1 | - | 2 | 455 | |||
BC016573.1 | - | 2 | 848 | |||
BC147770.1 | - | 2 | 558 | 9087493 | ||
BC147784.1 | - | 2 | 558 | 9087503 | ||
CK624100.1 | - | 2 | 344 | |||
DV655694.1 | - | 2 | 426 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
