siRNA Details
Gene Symbol: Aldh1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011921.2 | NP_036051.1 | 9 | 976 | ||
GenBank mRNA | BC032880.1 | - | 9 | 953 | ||
BC046315.1 | - | 9 | 950 | 6314273 | ||
BC139412.1 | - | 9 | 953 | 8862434 | ||
BC139413.1 | - | 9 | 953 | 8862435 | ||
U96401.1 | - | 9 | 976 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
