siRNA Details
Gene Symbol: Btd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025295.4 | NP_079571.1 | 4 | 602 | ||
GenBank mRNA | AK005506.1 | - | 4 | 595 | ||
AK135375.1 | - | 4 | 592 | |||
BC024051.1 | - | 4 | 569 | 5099802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
