siRNA Details
Gene Symbol: Cetn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007593.5 | NP_031619.3 | 1 | 112 | ||
GenBank mRNA | AK005736.1 | - | 1 | 91 | ||
AK005743.1 | - | 1 | 73 | |||
AK005837.1 | - | 1 | 113 | |||
AK006940.1 | - | 1 | 113 | |||
AK015808.1 | - | 111 | ||||
AK076986.1 | - | 122 | ||||
AK132753.1 | - | 1 | 112 | |||
BC048488.1 | - | 1 | 113 | 6775067 | ||
BC061155.1 | - | 1 | 111 | 6774850 | ||
BU961997.1 | - | 1 | 150 | |||
D16301.1 | - | 1 | 96 |

More Information
Other Species Targeted

Gene Ontology Categories:
