siRNA Details
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159500.1 | NP_001152972.1 | 5 | 1076 | ||
NM_011934.4 | NP_036064.3 | 5 | 975 | |||
GenBank mRNA | AK032025.1 | - | 5 | 783 | ||
AK044339.1 | - | 5 | 976 | |||
AK052256.1 | - | 5 | 1078 | |||
BC044858.1 | - | 4 | 645 | |||
BC132595.1 | - | 5 | 940 | 40130872 | ||
BC132597.1 | - | 5 | 941 | 40130874 | ||
S82458.1 | - | 4 | 683 | |||
X89594.1 | - | 4 | 683 |
