siRNA Details
Gene Symbol: Esrrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159500.1 | NP_001152972.1 | 6, 7 | 1498 | ||
NM_011934.4 | NP_036064.3 | 6, 7 | 1397 | |||
GenBank mRNA | AK032025.1 | - | 6, 7 | 1205 | ||
AK052256.1 | - | 6, 7 | 1500 | |||
BC044858.1 | - | 5, 6 | 1067 | |||
BC132595.1 | - | 6, 7 | 1362 | 40130872 | ||
BC132597.1 | - | 6, 7 | 1363 | 40130874 | ||
S82458.1 | - | 5, 6 | 1105 | |||
X89594.1 | - | 5, 6 | 1105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
