siRNA Details
Gene Symbol: Fto | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ237917.1 | - | 7 | 1192 | ||
AK088881.1 | - | 7 | 1192 | |||
AK129437.1 | - | 7 | 1173 | |||
AK194211.1 | - | 2 | 106 | |||
BC022222.1 | - | 7 | 1157 | 4237261 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
