siRNA Details
Gene Symbol: Hsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256042.1 | NP_001242971.1 | 2 | 215 | ||
NM_011939.3 | NP_036069.1 | 2 | 215 | |||
GenBank mRNA | AB029349.1 | - | 2 | 147 | ||
AB029350.1 | - | 2 | 142 | |||
AF160966.1 | - | 2 | 142 | |||
BC138130.1 | - | 2 | 215 | 8861149 | ||
BC145153.1 | - | 2 | 215 | |||
BC145154.1 | - | 2 | 215 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
