siRNA Details
Gene Symbol: Ifi202b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008327.2 | NP_032353.2 | 5 | 1176 | ||
NM_011940.2 | NP_036070.2 | 5 | 1173 | |||
GenBank mRNA | AF140672.1 | - | 5 | 1046 | ||
BC018233.1 | - | 1172 | 3990226 | |||
BC055888.1 | - | 4 | 1165 | 4945964 | ||
DQ222946.1 | - | 4 | 952 | |||
JX945582.1 | - | 4 | 1198 | |||
M31418.1 | - | 1245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
