siRNA Details
Gene Symbol: Ifi202b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF140672.1 | - | 5 | 983 | ||
BC018233.1 | - | 1109 | 3990226 | |||
BC055888.1 | - | 4 | 1102 | 4945964 | ||
DQ222946.1 | - | 4 | 889 | |||
JX945582.1 | - | 4 | 1135 | |||
M31418.1 | - | 1182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
