siRNA Details
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 11 | 1803 | ||
NM_009582.4 | NP_033608.3 | 9 | 1627 | |||
GenBank mRNA | AK029882.1 | - | 9 | 1436 | ||
AK034374.1 | - | 11 | 1619 | |||
AK081623.1 | - | 4 | 295 | |||
AK166435.1 | - | 11 | 1723 | |||
BC047158.1 | - | 8 | 1421 | 6494216 | ||
BC057572.1 | - | 11 | 1627 | 6401321 | ||
U14636.1 | - | 9 | 1459 | |||
U23789.1 | - | 8 | 1417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
