siRNA Details
Gene Symbol: Map3k5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008580.4 | NP_032606.4 | 2 | 655 | ||
GenBank mRNA | AB006787.3 | - | 2 | 707 | ||
BC116627.1 | - | 2 | 655 | 40086293 | ||
BC116628.1 | - | 2 | 655 | 40086299 | ||
BC133697.1 | - | 2 | 655 | 40086283 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
