siRNA Details
Gene Symbol: Map3k7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009316.1 | NP_033342.1 | 16 | 1725 | ||
NM_172688.3 | NP_766276.1 | 15 | 1644 | |||
GenBank mRNA | AK046565.1 | - | 15 | 1610 | ||
AK053911.1 | - | 15 | 1630 | |||
AK133642.1 | - | 9 | 771 | |||
AK137046.1 | - | 15 | 1645 | |||
AK159277.1 | - | 15 | 1608 | |||
CV559615.1 | - | 9 | 781 |
