siRNA Details
Gene Symbol: Prl2c4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011954.2 | NP_036084.2 | 804 | |||
GenBank mRNA | BC058816.1 | - | 804 | 30053850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
