siRNA Details
Gene Symbol: Orc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011958.3 | NP_036088.3 | 14 | 1305 | ||
GenBank mRNA | AJ003140.1 | - | 14 | 1266 | ||
AK010258.1 | - | 14 | 1289 | |||
AK011550.1 | - | 14 | 1219 | |||
AK076324.1 | - | 14 | 1306 | |||
AK084326.1 | - | 14 | 1274 | |||
AK145454.1 | - | 14 | 1317 | |||
AK169291.1 | - | 14 | 1289 | |||
BC015072.1 | - | 14 | 1277 | 4212266 | ||
BC019748.1 | - | 14 | 1247 | 5007346 | ||
Y16386.1 | - | 13 | 1137 |
