siRNA Details
Gene Symbol: Orc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011959.2 | NP_036089.1 | 4 | 460 | ||
GenBank mRNA | AJ007360.1 | - | 4 | 457 | ||
AK084683.1 | - | 4 | 488 | |||
AK144983.1 | - | 4 | 476 | |||
AK165912.1 | - | 4 | 466 | |||
AK166537.1 | - | 4 | 399 | |||
AK169683.1 | - | 4 | 434 | |||
BC006927.1 | - | 4 | 476 | 3601908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
