siRNA Details
Gene Symbol: Plod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142916.1 | NP_001136388.1 | 17, 18 | 2087 | ||
NM_011961.3 | NP_036091.2 | 16, 17 | 2024 | |||
GenBank mRNA | AF080572.1 | - | 16, 17 | 1795 | ||
AK033363.1 | - | 17, 18 | 2046 | |||
AK046364.1 | - | 16, 17 | 2028 | |||
AK157207.1 | - | 17, 18 | 2047 | |||
BC021352.1 | - | 16, 17 | 1856 | 5007509 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
